Assay Details
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss | ZNF705E MIR3165 LRTOMT FAM86C1 LOC100133315 DEFB108B LOC100128494 ALG1L9P RNF121 LOC100129216 IL18BP NUMA1 |
nsv951037 | Chr.11:72000055 - 72016354 on Build GRCh38 | Deletion | LOC100128494 IL18BP NUMA1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |