Assay Details
Target Gene Details
Entrez Gene ID: | 23162 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 3 |
Gene Aliases: |
JIP-3, JIP3, JSAP1, SYD2, syd |
Location: |
Chr.16:1706183-1770317 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK8IP3 | NM_001040439.1 | NP_001035529.1 | ||
NM_001318852.1 | NP_001305781.1 | |||
NM_015133.4 | NP_055948.2 | |||
XM_005255190.2 | XP_005255247.1 | |||
XM_006720869.2 | XP_006720932.1 | |||
XM_011522429.2 | XP_011520731.1 | |||
XM_011522430.2 | XP_011520732.1 | |||
XM_011522431.2 | XP_011520733.1 | |||
XM_011522432.2 | XP_011520734.1 | |||
XM_017023079.1 | XP_016878568.1 | |||
XM_017023080.1 | XP_016878569.1 | |||
XM_017023081.1 | XP_016878570.1 | |||
AB028989.2 | ||||
AB621811.1 | ||||
AK024437.1 | BAB15727.1 | |||
BC137123.1 | ||||
BC137124.1 | ||||
BC144486.1 | ||||
BC150266.1 | ||||
BY795428.2 | ||||
CN405915.1 | ||||
CR749696.1 | CAH18476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4856n54 | Chr.16:1600945 - 1813519 on Build GRCh38 | Loss | MIR3177 MAPK8IP3 IGFALS HN1L IFT140 HAGH MRPS34 EME2 CRAMP1 LOC105371046 NUBP2 SPSB3 NME3 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | FAM173A FLJ42627 UNKL MLST8 GFER PRSS27 IFT140 MPG LOC107987233 HAGHL LOC729652 MIR5587 CHTF18 C1QTNF8 LOC107984876 MIR3176 TRAF7 FAHD1 CLCN7 LINC00235 METTL26 NHLRC4 MAPK8IP3 SSTR5 AXIN1 MIR6767 LINC00254 NARFL SNORA64 MIR6768 NPW ERVK13-1 ARHGDIG BAIAP3 CACNA1H MIR4516 LOC106660606 DECR2 LUC7L RHBDF1 LOC105371047 MRPS34 BRICD5 NTN3 NTHL1 STUB1 TMEM204 RAB11FIP3 UBE2I METRN PGP SNRNP25 IGFALS TBC1D24 NPRL3 SNORA78 HN1L MEIOB ZNF598 HBA2 LMF1-AS1 JMJD8 RPS2 RNPS1 C16orf91 KCTD5 MRPL28 DDX11L10 LOC101929280 WDR90 SPSB3 SOX8 RPL23AP5 TPSG1 LOC100287175 TBL3 RHOT2 TPSD1 HBQ1 SNORA10 RPL3L MIR940 LOC652276 FAM234A HBA1 WDR24 C16orf59 LOC105371045 RAB26 NOXO1 MSLN LMF1 TSR3 DNASE1L2 POLR3K PRSS29P MIR3177 CEMP1 HBM SNHG19 SSTR5-AS1 TELO2 HBZ LOC100134368 CASKIN1 ECI1 LOC105371184 PTX4 TMEM8A RNF151 LOC105371046 SNORD60 SLC9A3R2 AMDHD2 MIR3180-5 WASIR2 WFIKKN1 MIR6859-4 CCDC78 PRR35 FBXL16 MCRIP2 PDPK1 MIR4717 MIR3178 RHBDL1 MIR662 LOC105371038 EME2 GNPTG CRAMP1 SNHG9 PDIA2 SYNGR3 RGS11 ABCA3 TSC2 PRR25 RAB40C NUBP2 PIGQ E4F1 NME3 NDUFB10 ATP6V0C RPUSD1 CCDC154 HS3ST6 CAPN15 TPSB2 TPSAB1 NME4 HAGH ABCA17P GNG13 MSRB1 MIR3677 LOC105371049 CCNF MIR6511B1 MIR1225 PKD1 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | MIR3177 FAHD1 MAPK8IP3 IGFALS TELO2 HN1L MEIOB IFT140 HAGH LOC105371047 MRPS34 EME2 CRAMP1 LOC105371046 NUBP2 SPSB3 TMEM204 NME3 |
nsv1160328 | Chr.16:1704619 - 1797867 on Build GRCh38 | Deletion | MRPS34 MIR3177 EME2 MAPK8IP3 IGFALS NUBP2 SPSB3 NME3 |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MIR3177 FAHD1 MAPK8IP3 IGFALS HN1L MEIOB HAGH MRPS34 EME2 CRAMP1 NUBP2 SPSB3 NME3 |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion | MRPS34 MIR3177 EME2 FAHD1 MAPK8IP3 IGFALS NUBP2 SPSB3 HAGH NME3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |