Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | 6 | 1625 | NP_001139528.1 |
NM_001146083.2 | 7 | 1704 | NP_001139555.1 | |
NM_024298.4 | 8 | 1831 | NP_077274.3 | |
XM_011527299.2 | 8 | 2035 | XP_011525601.1 | |
XM_011527300.2 | 8 | 1426 | XP_011525602.1 | |
AF211969.1 | 2 | 286 | ||
AK055908.1 | 8 | 1764 | BAB71043.1 | |
AK122789.1 | 9 | 1549 | ||
AK123290.1 | 2 | 2278 | ||
AK293199.1 | 6 | 1235 | ||
AK298689.1 | 6 | 1311 | ||
AK311493.1 | 7 | 1107 | ||
BC002512.2 | 7 | 1281 | AAH02512.2 | |
BC003164.1 | 8 | 1362 | AAH03164.2 | |
BC015857.1 | 7 | 1268 | ||
EU016381.1 | 7 | 1283 | ||
S82470.1 | 6 | 1137 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | MBOAT7 TSEN34 TMC4 LENG1 |
nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss | TFPT MBOAT7 NDUFA3 TMC4 LENG1 PRPF31 CNOT3 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | TFPT MBOAT7 OSCAR NDUFA3 TSEN34 TMC4 LENG1 PRPF31 CNOT3 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR516A2 MIR517C ZNF331 MIR518B MBOAT7 MIR516A1 OSCAR LILRB3 MIR371A MIR519D MIR520B LILRA5 TSEN34 VN1R4 FAM90A27P MIR1323 CACNG8 MIR527 MIR519B MIR520D ZNF818P MIR935 MIR520H MIR518F TARM1 NDUFA3 MIR524 MIR498 PRPF31 MIR512-2 DPRX MIR519E ZNF845 MIR525 VN1R2 MIR523 ZNF761 MIR512-1 BIRC8 LOC284379 MIR526B ZNF525 MIR1283-1 ZNF813 CNOT3 MIR520E CACNG7 MYADM CACNG6 MIR371B MIR518A1 MIR515-1 TFPT MIR520C MIR516B1 MIR516B2 MIR521-1 NLRP12 LENG1 MIR518E MIR373 LILRB5 PRKCG ZNF765 LILRB2 MIR521-2 MIR519C MIR517B MIR1283-2 MIR526A1 MIR518D MIR517A MIR520F TMC4 MIR372 MIR519A2 MIR518A2 LILRA6 ZNF677 MIR522 MIR519A1 MIR518C ZNF665 MIR520A MIR520G RPS9 LOC105372457 MIR526A2 MIR4752 MIR515-2 TPM3P9 VSTM1 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | MBOAT7 LILRB3 RPS9 TSEN34 TMC4 LILRB5 LENG1 LILRA6 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | KIR3DL1 LAIR2 LOC101928804 MBOAT7 KIR3DL2 LILRA4 LILRB3 LILRA5 TSEN34 KIR2DS4 NLRP7 CDC42EP5 LENG9 FCAR KIR3DX1 LENG8-AS1 LILRB5 TTYH1 LILRB2 LAIR1 KIR2DL3 TMC4 LILRA2 LILRA6 LILRA1 NCR1 KIR2DL4 MIR8061 KIR3DL3 LILRB4 RPS9 LILRB1 KIR2DL1 LENG8 MIR4752 LOC102724300 LILRP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |