Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | 5 | 699 | NP_001028285.1 |
NM_198585.2 | 5 | 699 | NP_940987.2 | |
XM_011518665.2 | 6 | 1005 | XP_011516967.1 | |
XM_011518666.2 | 6 | 1015 | XP_011516968.1 | |
XM_011518668.1 | 3 | 360 | XP_011516970.1 | |
AY359088.1 | 5 | 699 | AAQ89446.1 | |
AY430414.1 | 4 | 532 | AAR04374.1 | |
AY903953.1 | 3 | 645 | AAW83515.1 | |
AY903954.1 | 4 | 515 | AAW83516.1 | |
BC141810.1 | 5 | 600 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss | MIR7114 ENTPD8 NSMF |
nsv1049808 | Chr.9:137356366 - 137438026 on Build GRCh38 | Gain | NOXA1 ENTPD8 EXD3 |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion | MIR7114 NOXA1 ENTPD8 EXD3 NSMF PNPLA7 |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion | NOXA1 ENTPD8 EXD3 |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | NOXA1 ENTPD8 EXD3 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | TRAF2 MAN1B1 PRR31 NDOR1 EDF1 TUBB4B LCN12 LRRC26 SAPCD2 STPG3 C9orf172 SLC34A3 ENTPD8 UAP1L1 MIR7114 CCDC183-AS1 TMEM203 TMEM210 RABL6 RNF224 MAMDC4 PTGDS ENTPD2 MAN1B1-AS1 NSMF MIR4292 PNPLA7 PHPT1 CLIC3 FBXW5 GRIN1 FAM166A ABCA2 NPDC1 DPP7 CCDC183 MIR4479 C9orf142 TOR4A C9orf139 NELFB NRARP MIR3621 C8G LCNL1 ANAPC2 NOXA1 SSNA1 FUT7 LOC101930307 C9orf173-AS1 TPRN RNF208 EXD3 CYSRT1 |
esv3622034 | Chr.9:137433077 - 137439026 on Build GRCh38 | Gain | NOXA1 ENTPD8 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | RNF224 STPG3 SLC34A3 ENTPD8 TOR4A NELFB NRARP MIR3621 ANAPC2 NDOR1 NOXA1 SSNA1 GRIN1 C9orf173-AS1 FAM166A TMEM203 TUBB4B TPRN RNF208 EXD3 TMEM210 CYSRT1 LRRC26 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |