Assay Details
Target Gene Details
Entrez Gene ID: | 29074 |
Gene Name: | mitochondrial ribosomal protein L18 |
Gene Aliases: |
HSPC071, L18mt, MRP-L18 |
Location: |
Chr.6:159789812-159798429 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL18 | NM_001318817.1 | 4 | 589 | NP_001305746.1 |
NM_014161.4 | 4 | 738 | NP_054880.2 | |
NR_134860.1 | 5 | 858 | ||
NR_134861.1 | 4 | 815 | ||
NR_134862.1 | 4 | 738 | ||
NR_134863.1 | 4 | 722 | ||
NR_134864.1 | 4 | 621 | ||
NR_134865.1 | 4 | 589 | ||
NR_134866.1 | 4 | 567 | ||
AF161556.1 | 4 | 716 | AAF29043.1 | |
AK311391.1 | 5 | 735 | ||
AL136633.1 | 4 | 688 | CAB66568.1 | |
AV756740.1 | 4 | 524 | ||
BC001623.1 | 4 | 709 | AAH01623.1 | |
BF797811.1 | 4 | 622 | ||
BG548185.1 | 4 | 569 | ||
BM924328.1 | 4 | 831 | ||
BU601065.1 | 4 | 679 | ||
BU932963.1 | 4 | 727 | ||
CA777760.1 | 4 | 449 | ||
CB529016.1 | 1 | 271 | ||
CD675657.1 | ||||
CR533491.1 | CAG38522.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |