Assay Details
Target Gene Details
Entrez Gene ID: | 8503 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 3 |
Gene Aliases: |
p55, p55-GAMMA, p55PIK |
Location: |
Chr.1:46040140-46176495 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3R3 | NM_001114172.1 | 11 | 4772 | NP_001107644.1 |
NM_001303427.1 | 10 | 4677 | NP_001290356.1 | |
NM_001303428.1 | 11 | 4616 | NP_001290357.1 | |
NM_001303429.1 | 9 | 5094 | NP_001290358.1 | |
NM_003629.3 | 10 | 5271 | NP_003620.3 | |
XM_017002614.1 | 11 | 4649 | XP_016858103.1 | |
BC021622.1 | 10 | 5205 | AAH21622.1 | |
BX537388.1 | 11 | 4616 | CAD97630.1 | |
CR749649.1 | 10 | 5192 | CAH18443.1 | |
U90907.1 | 1 | 1253 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829737 | Chr.1:45887228 - 46040752 on Build GRCh38 | Gain | MAST2 PIK3R3 |
nsv1007750 | Chr.1:46005621 - 46086892 on Build GRCh38 | Gain | MAST2 PIK3R3 |
esv3577805 | Chr.1:46003363 - 46075520 on Build GRCh38 | Gain | MAST2 PIK3R3 |
dgv31e212 | Chr.1:45920597 - 46063492 on Build GRCh38 | Gain | MAST2 PIK3R3 |
esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain | NSUN4 LOC101929626 RAD54L UQCRH FAAHP1 KNCN MKNK1-AS1 TSPAN1 LURAP1 MKNK1 LINC01398 LRRC41 MAST2 FAAH DMBX1 PIK3R3 POMGNT1 |
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | TESK2 MMACHC CCDC163P NASP TMEM69 IPP CCDC17 PRDX1 RPS15AP10 MAST2 AKR1A1 GPBP1L1 PIK3R3 |
dgv19e214 | Chr.1:46030708 - 46154212 on Build GRCh38 | Gain | LOC101929626 MAST2 PIK3R3 |
esv3585925 | Chr.1:45992755 - 46084727 on Build GRCh38 | Gain | MAST2 PIK3R3 |
nsv1009527 | Chr.1:45820657 - 46054455 on Build GRCh38 | Gain | MAST2 PIK3R3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |