Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC100129503 ACTG1 AATK-AS1 TEPSIN CCDC137 FSCN2 LOC105371925 CEP131 LOC101928855 BAIAP2 TSPAN10 MRPL12 MIR3065 C17orf89 MIR1250 BAHCC1 LINC00482 CHMP6 MIR3186 OXLD1 MIR6786 RPTOR MIR4740 SLC38A10 BAIAP2-AS1 FAAP100 ARL16 LOC107985021 LOC107985022 HGS SLC25A10 LOC105371921 TMEM105 NPLOC4 LOC400627 AATK MIR657 PDE6G MIR338 LOC100130370 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK-AS1 AATK CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | HEXDC LOC105371944 SECTM1 AATK-AS1 CCDC137 FSCN2 TBCD CEP131 RFNG FN3K NPB PYCR1 OGFOD3 P4HB GPS1 LOC101929511 LINC00482 CSNK1D SNORD134 DCXR SLC16A3 FOXK2 MIR3186 LRRC45 MIR6786 LOC105376790 NOTUM DUS1L FAAP100 CD7 MIR6787 ARL16 CCDC57 HGS ASPSCR1 SLC25A10 LOC101929552 AATK MAFG-AS1 TEX19 PDE6G ACTG1 MYADML2 TEPSIN MAFG LOC105371925 MIR4525 TSPAN10 PCYT2 NARF PPP1R27 MRPL12 C17orf62 WDR45B C17orf89 BAHCC1 GCGR ALYREF OXLD1 ANAPC11 STRA13 UTS2R MIR4740 SLC38A10 ZNF750 B3GNTL1 LOC105376791 RAB40B ARHGDIA SIRT7 TMEM105 NPLOC4 MCRIP1 FN3KRP FASN LOC100130370 RAC3 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | AATK-AS1 C17orf89 TEPSIN SLC38A10 AATK LOC105371925 CEP131 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR3065 AATK-AS1 MIR1250 AATK MIR657 CEP131 MIR338 BAIAP2 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | AATK-AS1 TEPSIN SLC38A10 BAIAP2-AS1 LOC105371925 CEP131 BAIAP2 MIR3065 TMEM105 C17orf89 MIR1250 AATK LINC00482 MIR657 MIR338 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR3065 AATK-AS1 MIR1250 AATK MIR657 BAIAP2-AS1 CEP131 MIR338 BAIAP2 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | AATK-AS1 C17orf89 TEPSIN SLC38A10 LOC105371925 CEP131 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | MIR3186 ACTG1 AATK-AS1 MIR4740 TEPSIN SLC38A10 FSCN2 LOC105371925 FAAP100 CEP131 TMEM105 NPLOC4 C17orf89 AATK BAHCC1 LINC00482 LOC100130370 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | MIR3065 AATK-AS1 MIR1250 AATK MIR657 MIR338 BAIAP2 |