Assay Details
Target Gene Details
Entrez Gene ID: | 10297 |
Gene Name: | APC2, WNT signaling pathway regulator |
Gene Aliases: |
APCL |
Location: |
Chr.19:1446268-1473244 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APC2 | NM_005883.2 | NP_005874.1 | ||
XM_005259475.2 | XP_005259532.1 | |||
XM_006722607.2 | XP_006722670.1 | |||
XM_006722608.3 | XP_006722671.2 | |||
XM_006722609.3 | XP_006722672.1 | |||
XM_006722610.3 | XP_006722673.2 | |||
AB012162.1 | BAA34611.1 | |||
AF110334.1 | AAD28183.1 | |||
AF128222.1 | AAF01784.1 | |||
AJ012652.1 | CAB61207.1 | |||
AL078616.1 | ||||
BC032573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | REXO1 SCAMP4 CSNK1G2-AS1 MBD3 ABHD17A BTBD2 ONECUT3 ADAT3 ATP8B3 NDUFS7 RPS15 APC2 GAMT C19orf25 KLF16 DAZAP1 ADAMTSL5 MEX3D CSNK1G2 UQCR11 MIR1909 MUM1 LOC100288123 PLK5 REEP6 TCF3 PCSK4 |
nsv578279 | Chr.19:1413575 - 1530061 on Build GRCh38 | Gain | PLK5 REEP6 APC2 C19orf25 PCSK4 DAZAP1 ADAMTSL5 RPS15 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | PLK5 REEP6 APC2 GAMT C19orf25 PCSK4 DAZAP1 NDUFS7 ADAMTSL5 RPS15 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | POLR2E MIDN EFNA2 GPX4 CIRBP-AS1 TMEM259 NDUFS7 RPS15 CBARP LOC102725180 APC2 ABCA7 GAMT C19orf25 STK11 ATP5D DAZAP1 ADAMTSL5 WDR18 CIRBP GRIN3B C19orf24 SBNO2 RNU6-2 ARHGAP45 ARID3A MUM1 CNN2 PLK5 REEP6 LOC105372235 PCSK4 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | REXO1 SCAMP4 CSNK1G2-AS1 MBD3 ABHD17A BTBD2 ONECUT3 ADAT3 ATP8B3 NDUFS7 RPS15 APC2 GAMT C19orf25 KLF16 DAZAP1 ADAMTSL5 MEX3D CSNK1G2 UQCR11 MKNK2 MIR1909 MUM1 LOC100288123 PLK5 REEP6 TCF3 PCSK4 |
nsv470105 | Chr.19:1444420 - 1526614 on Build GRCh38 | Gain | PLK5 REEP6 APC2 C19orf25 PCSK4 ADAMTSL5 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |