Assay Details
Target Gene Details
Entrez Gene ID: | 1937 |
Gene Name: | eukaryotic translation elongation factor 1 gamma |
Gene Aliases: |
EF1G, GIG35 |
Location: |
Chr.11:62559601-62573988 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EEF1G | NM_001404.4 | NP_001395.1 | ||
AK092787.1 | ||||
AK129569.1 | ||||
AK129618.1 | ||||
AK130026.1 | ||||
AK299876.1 | ||||
AK300203.1 | ||||
AK300686.1 | ||||
AK300729.1 | ||||
AL556034.3 | ||||
AY542307.1 | AAT08176.1 | |||
AY687925.1 | ||||
BQ947806.1 | ||||
M55409.1 | AAC18414.1 | |||
X63526.1 | CAA45089.1 | |||
Z11531.1 | CAA77630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | EEF1G TUT1 ROM1 AHNAK B3GAT3 MTA2 EML3 GANAB MIR6747 |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | EEF1G TUT1 AHNAK MTA2 MIR6747 |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | EEF1G TUT1 ROM1 B3GAT3 MTA2 EML3 GANAB MIR6747 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | LRRN4CL INTS5 ROM1 BSCL2 C11orf98 HNRNPUL2-BSCL2 UQCC3 SNORA57 LBHD1 GANAB UBXN1 MIR6747 EEF1G TUT1 AHNAK B3GAT3 MTA2 METTL12 EML3 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | HNRNPUL2 GNG3 LRRN4CL INTS5 ROM1 BSCL2 C11orf98 TTC9C HNRNPUL2-BSCL2 UQCC3 SNORA57 LBHD1 GANAB UBXN1 MIR6747 EEF1G TUT1 B3GAT3 MTA2 METTL12 EML3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |