Assay Details
Target Gene Details
Entrez Gene ID: | 7536 |
Gene Name: | splicing factor 1 |
Gene Aliases: |
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162 |
Location: |
Chr.11:64764604-64779043 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469962 | Chr.11:64759608 - 64854101 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv225n67 | Chr.11:64770620 - 64783455 on Build GRCh38 | Gain |
![]() |
dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv153n27 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)