Assay Details
Target Gene Details
Entrez Gene ID: | 92799 |
Gene Name: | SH3KBP1 binding protein 1 |
Gene Aliases: |
PP203, Sb1 |
Location: |
Chr.19:40576851-40591399 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHKBP1 | NM_138392.3 | NP_612401.2 | ||
XM_006723474.1 | XP_006723537.1 | |||
XM_006723475.1 | XP_006723538.1 | |||
XM_011527492.1 | XP_011525794.1 | |||
XM_011527493.1 | XP_011525795.1 | |||
XM_017027473.1 | XP_016882962.1 | |||
XM_017027474.1 | XP_016882963.1 | |||
AF258553.1 | AAG23756.1 | |||
AK075057.1 | BAC11374.1 | |||
AK123695.1 | ||||
AK297006.1 | ||||
AK300808.1 | ||||
AK300819.1 | ||||
AK312748.1 | ||||
AY007117.1 | ||||
BC007653.1 | AAH07653.1 | |||
BC022855.1 | AAH22855.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |