Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | NP_064536.2 | ||
XM_011511499.1 | XP_011509801.1 | |||
AF270647.1 | AAF81750.1 | |||
AK054615.1 | ||||
AK291190.1 | ||||
AL542720.3 | ||||
BC007894.2 | AAH07894.1 | |||
BC008894.2 | AAH08894.1 | |||
BC009998.2 | AAH09998.1 | |||
BC012774.2 | AAH12774.1 | |||
BC012793.2 | AAH12793.1 | |||
BC013279.2 | AAH13279.1 | |||
BC014076.2 | AAH14076.1 | |||
BC014274.2 | AAH14274.3 | |||
BC032106.1 | AAH32106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | STARD7 TMEM127 ITPRIPL1 SNRNP200 NCAPH CIAO1 LOC105373495 STARD7-AS1 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | ANKRD36C LOC100506123 ANKRD36 ADRA2B KANSL3 FAHD2CP ASTL ANKRD39 LOC100506076 SNRNP200 LOC653924 LMAN2L GPAT2 ARID5A FER1L5 LOC107984110 LOC105373495 FAHD2B STARD7-AS1 SEMA4C DUSP2 CNNM3 NCAPH LOC105373496 NEURL3 CNNM4 ANKRD23 MIR3127 STARD7 TMEM127 LOC101927053 ANKRD36B ITPRIPL1 CIAO1 FAM178B |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | ANKRD36C LOC100506123 ANKRD36 ADRA2B KANSL3 FAHD2CP ASTL ANKRD39 LOC100506076 SNRNP200 LOC653924 LMAN2L GPAT2 ARID5A FER1L5 LOC107984110 LOC105373495 FAHD2B STARD7-AS1 SEMA4C DUSP2 CNNM3 NCAPH LOC105373496 NEURL3 CNNM4 ANKRD23 MIR3127 STARD7 TMEM127 LOC101927053 ANKRD36B ITPRIPL1 CIAO1 FAM178B |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | ANKRD36C LOC100506123 ANKRD36 ADRA2B KANSL3 FAHD2CP ASTL ANKRD39 LOC100506076 SNRNP200 LOC653924 LMAN2L GPAT2 ARID5A FER1L5 LOC107984110 LOC105373495 FAHD2B STARD7-AS1 SEMA4C DUSP2 CNNM3 NCAPH LOC105373496 NEURL3 CNNM4 ANKRD23 MIR3127 STARD7 TMEM127 LOC101927053 ANKRD36B ITPRIPL1 CIAO1 FAM178B |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |