Assay Details
Target Gene Details
Entrez Gene ID: | 4978 |
Gene Name: | opioid binding protein/cell adhesion molecule like |
Gene Aliases: |
IGLON1, OBCAM, OPCM |
Location: |
Chr.11:132403361-133532983 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OPCML | NM_001012393.2 | 8 | 6506 | NP_001012393.1 |
NM_001319103.1 | 8 | 6811 | NP_001306032.1 | |
NM_001319104.1 | 7 | 6421 | NP_001306033.1 | |
NM_001319105.1 | 8 | 6330 | NP_001306034.1 | |
NM_001319106.1 | 7 | 6781 | NP_001306035.1 | |
NM_002545.4 | 7 | 6784 | NP_002536.1 | |
XM_006718846.2 | 8 | 6729 | XP_006718909.1 | |
XM_011542856.2 | 9 | 7042 | XP_011541158.1 | |
AF070577.1 | ||||
BX537377.1 | CAD97619.1 | |||
DA247864.1 | 1 | 259 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |