Assay Details
Target Gene Details
Entrez Gene ID: | 6347 |
Gene Name: | C-C motif chemokine ligand 2 |
Gene Aliases: |
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF |
Location: |
Chr.17:34255277-34257203 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL2 | NM_002982.3 | NP_002973.1 | ||
AK311960.1 | ||||
AV733621.2 | ||||
BC009716.1 | AAH09716.1 | |||
BG530064.1 | ||||
BT007329.1 | AAP35993.1 | |||
M24545.1 | AAA18164.1 | |||
M26683.1 | 1 | 147 | AAA96708.1 | |
M28226.1 | AAA60309.1 | |||
S69738.1 | AAB29926.1 | |||
S71513.1 | AAB20651.1 | |||
X14768.1 | CAA32876.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)