Assay Details
Target Gene Details
Entrez Gene ID: | 1917 |
Gene Name: | eukaryotic translation elongation factor 1 alpha 2 |
Gene Aliases: |
EEF1AL, EF-1-alpha-2, EF1A, EIEE33, HS1, MRD38, STN, STNL |
Location: |
Chr.20:63488012-63499315 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EEF1A2 | NM_001958.3 | NP_001949.1 | ||
AB209064.1 | BAD92301.1 | |||
AB451389.1 | ||||
AK307536.1 | ||||
BC000432.2 | AAH00432.1 | |||
BC110409.1 | ||||
L10340.1 | AAA91835.1 | |||
X70940.1 | CAA50280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv7666n54 | Chr.20:63460989 - 63528151 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv3448 | Chr.20:63488613 - 63520639 on Build GRCh38 | Insertion |
![]() |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv586568 | Chr.20:63486715 - 63502196 on Build GRCh38 | Loss |
![]() |
nsv834033 | Chr.20:63421436 - 63554987 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv834034 | Chr.20:63462825 - 63625901 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758806 | Chr.20:63470996 - 63745014 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)