Assay Details
Target Gene Details
Entrez Gene ID: | 201266 |
Gene Name: | solute carrier family 39 member 11 |
Gene Aliases: |
C17orf26, ZIP11 |
Location: |
Chr.17:72645946-73092714 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC39A11 | NM_001159770.1 | 10 | 1868 | NP_001153242.1 |
NM_139177.3 | 10 | 1847 | NP_631916.2 | |
XM_005257134.3 | 10 | 1934 | XP_005257191.1 | |
XM_006721754.3 | 10 | 1955 | XP_006721817.1 | |
XM_006721755.2 | 11 | 2159 | XP_006721818.1 | |
XM_006721757.2 | 9 | 1697 | XP_006721820.1 | |
XM_011524493.1 | 11 | 2250 | XP_011522795.1 | |
XM_011524494.2 | 10 | 1901 | XP_011522796.1 | |
XM_017024330.1 | 11 | 2138 | XP_016879819.1 | |
XM_017024331.1 | 11 | 2229 | XP_016879820.1 | |
XM_017024332.1 | 10 | 1844 | XP_016879821.1 | |
XM_017024333.1 | 9 | 1808 | XP_016879822.1 | |
XM_017024334.1 | 9 | 1784 | XP_016879823.1 | |
XM_017024335.1 | 9 | 1676 | XP_016879824.1 | |
XM_017024336.1 | 11 | 2048 | XP_016879825.1 | |
XM_017024337.1 | 11 | 1961 | XP_016879826.1 | |
AF331643.1 | 10 | 1809 | AAL32175.1 | |
AK095227.1 | 10 | 1842 | BAC04504.1 | |
BC035631.1 | 10 | 1826 | AAH35631.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |