Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 | ||||
DA945486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | E2F4 CTRL ATP6V0D1 HSD11B2 KCTD19 DPEP3 DDX28 CENPT ACD PSKH1 EDC4 TMEM208 FAM65A KIAA0895L PARD6A ELMO3 TPPP3 FBXL8 EXOC3L1 CTCF FHOD1 NUTF2 MIR328 RANBP10 DUS2 LOC105369155 C16orf86 LRRC36 LCAT LOC100505942 THAP11 ZDHHC1 TRADD DPEP2 NRN1L LRRC29 LOC100131303 ENKD1 TSNAXIP1 CARMIL2 SLC12A4 NOL3 PSMB10 SLC9A5 AGRP PLEKHG4 GFOD2 HSF4 LOC107984813 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | LRRC29 E2F4 LOC105369155 TMEM208 NOL3 KIAA0895L C16orf70 SLC9A5 ELMO3 B3GNT9 TRADD FBXL8 EXOC3L1 PLEKHG4 FHOD1 HSF4 MIR328 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | LRRC29 E2F4 LOC105369155 TMEM208 NOL3 KIAA0895L C16orf70 ELMO3 B3GNT9 TRADD FBXL8 EXOC3L1 FHOD1 HSF4 MIR328 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | LRRC29 E2F4 LOC105369155 ELMO3 FBXL8 NOL3 KIAA0895L EXOC3L1 HSF4 MIR328 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | LRRC29 SLC9A5 E2F4 LOC105369155 ELMO3 TMEM208 KIAA0895L EXOC3L1 PLEKHG4 FHOD1 MIR328 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | LRRC29 E2F4 LOC105369155 TMEM208 NOL3 KIAA0895L SLC9A5 ELMO3 KCTD19 EXOC3L1 PLEKHG4 FHOD1 MIR328 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | E2F4 CTRL ATP6V0D1 HSD11B2 KCTD19 DPEP3 DDX28 CENPT ACD PSKH1 EDC4 TMEM208 FAM65A KIAA0895L PARD6A C16orf70 ELMO3 TPPP3 FBXL8 EXOC3L1 CTCF FHOD1 NUTF2 MIR328 RANBP10 DUS2 LOC105369155 C16orf86 LRRC36 LCAT LOC100505942 THAP11 ZDHHC1 TRADD DPEP2 NRN1L LRRC29 LOC100131303 ENKD1 TSNAXIP1 CARMIL2 SLC12A4 NOL3 PSMB10 SLC9A5 AGRP B3GNT9 PLEKHG4 GFOD2 HSF4 LOC107984813 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |