Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | 6 | 926 | NP_001028285.1 |
NM_198585.2 | 6 | 926 | NP_940987.2 | |
XM_011518665.2 | 7 | 1232 | XP_011516967.1 | |
XM_011518666.2 | 7 | 1242 | XP_011516968.1 | |
XM_011518668.1 | 4 | 587 | XP_011516970.1 | |
AY359088.1 | 6 | 926 | AAQ89446.1 | |
AY430414.1 | 5 | 759 | AAR04374.1 | |
AY903953.1 | 3 | 989 | AAW83515.1 | |
AY903954.1 | 5 | 742 | AAW83516.1 | |
BC141810.1 | 6 | 827 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss | ENTPD8 NSMF MIR7114 |
nsv1049808 | Chr.9:137356366 - 137438026 on Build GRCh38 | Gain | EXD3 ENTPD8 NOXA1 |
esv2739446 | Chr.9:137436134 - 137436605 on Build GRCh38 | Deletion | ENTPD8 |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion | EXD3 PNPLA7 ENTPD8 NSMF NOXA1 MIR7114 |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion | EXD3 ENTPD8 NOXA1 |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | EXD3 ENTPD8 NOXA1 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | STPG3 UAP1L1 RABL6 DPP7 FAM166A CCDC183 C9orf173-AS1 NOXA1 TRAF2 MAN1B1-AS1 TMEM203 TUBB4B NRARP NELFB CYSRT1 NPDC1 TOR4A NDOR1 CLIC3 LCNL1 PTGDS EDF1 C8G MIR4479 ANAPC2 PNPLA7 MIR7114 TPRN SSNA1 MAN1B1 MIR4292 FBXW5 ABCA2 CCDC183-AS1 SLC34A3 RNF208 SAPCD2 NSMF MAMDC4 PHPT1 MIR3621 LRRC26 C9orf142 TMEM210 GRIN1 PRR31 C9orf172 ENTPD2 LCN12 C9orf139 EXD3 RNF224 ENTPD8 FUT7 LOC101930307 |
esv3622034 | Chr.9:137433077 - 137439026 on Build GRCh38 | Gain | ENTPD8 NOXA1 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | STPG3 ANAPC2 TUBB4B NRARP FAM166A TPRN SSNA1 MIR3621 LRRC26 TMEM210 NELFB GRIN1 CYSRT1 TOR4A NDOR1 EXD3 RNF224 ENTPD8 C9orf173-AS1 SLC34A3 NOXA1 RNF208 TMEM203 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |