Assay Details
Target Gene Details
Entrez Gene ID: | 83639 |
Gene Name: | testis expressed 101 |
Gene Aliases: |
CT131, GTPR867, NYD-SP8, PRO1884, SGRG, SPATA44, TES101RP |
Location: |
Chr.19:43388611-43418615 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TEX101 | NM_001130011.1 | NP_001123483.1 | ||
NM_031451.4 | NP_113639.4 | |||
XM_005259303.3 | XP_005259360.2 | |||
AF241268.1 | AAK28327.1 | |||
AF353396.1 | AAK38662.1 | |||
AY014285.1 | AAK27310.1 | |||
AY359077.1 | AAQ89436.1 | |||
BC001861.2 | AAH01861.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469666 | Chr.19:43272995 - 43430182 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1057949 | Chr.19:43380906 - 43431018 on Build GRCh38 | Loss |
![]() |
esv3583431 | Chr.19:43412826 - 43449282 on Build GRCh38 | Loss |
![]() |
esv3644466 | Chr.19:43383591 - 43418114 on Build GRCh38 | Gain |
![]() |
dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060280 | Chr.19:43362671 - 43455533 on Build GRCh38 | Gain |
![]() ![]() |
nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509744 | Chr.19:43400292 - 43476817 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
dgv3604n100 | Chr.19:43414688 - 43453235 on Build GRCh38 | Loss |
![]() |
nsv520852 | Chr.19:43380933 - 43455962 on Build GRCh38 | Gain+Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)