Assay Details
Target Gene Details
Entrez Gene ID: | 5099 |
Gene Name: | protocadherin 7 |
Gene Aliases: |
BH-Pcdh, BHPCDH, PPP1R120 |
Location: |
Chr.4:30720378-31146801 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)