Assay Details
Target Gene Details
Entrez Gene ID: | 8763 |
Gene Name: | CD164 molecule |
Gene Aliases: |
DFNA66, MGC-24, MUC-24, endolyn |
Location: |
Chr.6:109366514-109382559 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD164 | NM_001142401.1 | NP_001135873.1 | ||
NM_001142402.1 | NP_001135874.1 | |||
NM_001142403.1 | NP_001135875.1 | |||
NM_001142404.1 | NP_001135876.1 | |||
NM_006016.4 | NP_006007.2 | |||
AF106518.1 | AAC82473.1 | |||
AF263279.1 | AAF85965.1 | |||
AF299341.1 | AAG53906.1 | |||
AF299342.1 | AAG53907.1 | |||
AF299343.1 | AAG53908.1 | |||
AK298684.1 | ||||
AK301692.1 | ||||
AK303525.1 | ||||
AK307193.1 | ||||
AK308664.1 | ||||
AK310854.1 | ||||
AK312357.1 | ||||
AK315908.1 | ||||
BC011522.2 | AAH11522.1 | |||
D14043.1 | BAA03130.1 | |||
DC403839.1 | ||||
FJ200494.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |