Assay Details
Target Gene Details
Entrez Gene ID: | 283337 |
Gene Name: | zinc finger protein 740 |
Gene Aliases: |
Zfp740 |
Location: |
Chr.12:53180715-53190870 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF740 | NM_001004304.3 | 7 | 1291 | NP_001004304.1 |
XM_006719344.2 | XP_006719407.1 | |||
XM_011538194.2 | XP_011536496.1 | |||
XM_017019181.1 | 7 | 1525 | XP_016874670.1 | |
AK091946.1 | 4 | 674 | ||
BC053557.1 | 7 | 1294 | AAH53557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | ZNF740 RARG MFSD5 IGFBP6 CSAD SOAT2 ITGB7 SPRYD3 |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | ZNF740 ESPL1 RARG MFSD5 CSAD SOAT2 ITGB7 |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | ZNF740 RARG IGFBP6 CSAD SOAT2 ITGB7 |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | EIF4B ESPL1 PFDN5 RARG C12orf10 AAAS SOAT2 SP7 ZNF740 MFSD5 IGFBP6 MIR6757 CSAD LOC283335 TNS2 ITGB7 SPRYD3 |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | ZNF740 RARG CSAD SOAT2 ITGB7 |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | ZNF740 RARG IGFBP6 CSAD TNS2 SOAT2 ITGB7 SPRYD3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |