Assay Details
Target Gene Details
Entrez Gene ID: | 3299 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
CTM, CTRCT5 |
Location: |
Chr.16:67163385-67169945 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSF4 | NM_001040667.2 | NP_001035757.1 | ||
NM_001538.3 | NP_001529.2 | |||
AB029348.1 | BAA84582.1 | |||
AK225152.1 | ||||
AK296210.1 | ||||
BM768246.1 | ||||
CA412687.1 | ||||
D87673.1 | BAA13433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | ACD DPEP3 PARD6A THAP11 EDC4 ENKD1 GFOD2 KIAA0895L DUS2 AGRP DPEP2 TMEM208 FBXL8 ZDHHC1 PSKH1 ELMO3 HSD11B2 E2F4 NOL3 MIR328 RANBP10 ATP6V0D1 HSF4 NRN1L LOC107984813 FHOD1 LRRC29 LRRC36 CARMIL2 FAM65A LOC100131303 PLEKHG4 NUTF2 TRADD SLC9A5 DDX28 LOC105369155 LOC100505942 CTRL CTCF C16orf86 KCTD19 EXOC3L1 PSMB10 SLC12A4 LCAT CENPT TPPP3 TSNAXIP1 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | TMEM208 HSF4 C16orf70 LOC105369155 FBXL8 ELMO3 KIAA0895L FHOD1 LRRC29 B3GNT9 E2F4 NOL3 EXOC3L1 PLEKHG4 TRADD MIR328 SLC9A5 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | TMEM208 HSF4 C16orf70 LOC105369155 FBXL8 ELMO3 KIAA0895L FHOD1 LRRC29 B3GNT9 E2F4 NOL3 EXOC3L1 TRADD MIR328 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | KIAA0895L HSF4 LRRC29 E2F4 LOC105369155 NOL3 EXOC3L1 FBXL8 MIR328 ELMO3 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | ACD DPEP3 PARD6A THAP11 EDC4 ENKD1 GFOD2 KIAA0895L DUS2 AGRP DPEP2 TMEM208 FBXL8 ZDHHC1 PSKH1 ELMO3 HSD11B2 E2F4 NOL3 MIR328 RANBP10 ATP6V0D1 HSF4 NRN1L LOC107984813 FHOD1 LRRC29 LRRC36 CARMIL2 FAM65A LOC100131303 PLEKHG4 NUTF2 TRADD SLC9A5 DDX28 C16orf70 LOC105369155 LOC100505942 CTRL CTCF B3GNT9 C16orf86 KCTD19 EXOC3L1 PSMB10 SLC12A4 LCAT CENPT TPPP3 TSNAXIP1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |