Assay Details
Target Gene Details
Entrez Gene ID: | 57489 |
Gene Name: | outer dense fiber of sperm tails 2 like |
Gene Aliases: |
dJ977L11.1 |
Location: |
Chr.1:86346824-86396345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ODF2L | NM_001007022.2 | NP_001007023.2 | ||
NM_001184765.1 | NP_001171694.1 | |||
NM_001184766.1 | NP_001171695.1 | |||
NM_020729.2 | NP_065780.2 | |||
XM_005271054.3 | XP_005271111.1 | |||
XM_011541815.2 | XP_011540117.1 | |||
XM_011541819.2 | XP_011540121.1 | |||
XM_011541821.2 | XP_011540123.1 | |||
XM_017001876.1 | XP_016857365.1 | |||
XM_017001877.1 | XP_016857366.1 | |||
XM_017001878.1 | XP_016857367.1 | |||
XM_017001879.1 | XP_016857368.1 | |||
XM_017001880.1 | XP_016857369.1 | |||
XM_017001881.1 | XP_016857370.1 | |||
XM_017001882.1 | XP_016857371.1 | |||
AB033055.1 | ||||
AK302792.1 | ||||
AK303209.1 | ||||
BC047368.1 | AAH47368.1 | |||
BC091490.1 | AAH91490.1 | |||
BM971230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546722 | Chr.1:85812632 - 86425390 on Build GRCh38 | Loss | COL24A1 MIR7856 CLCA2 ODF2L |
nsv1002541 | Chr.1:86223405 - 86440219 on Build GRCh38 | Gain | MIR7856 CLCA2 ODF2L |