Assay Details
Target Gene Details
Entrez Gene ID: | 3579 |
Gene Name: | C-X-C motif chemokine receptor 2 |
Gene Aliases: |
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB |
Location: |
Chr.2:218125290-218137253 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCR2 | NM_001168298.1 | 4 | 713 | NP_001161770.1 |
NM_001557.3 | 3 | 921 | NP_001548.1 | |
XM_005246530.3 | 4 | 693 | XP_005246587.1 | |
XM_017003990.1 | 4 | 673 | XP_016859479.1 | |
XM_017003991.1 | 2 | 584 | XP_016859480.1 | |
XM_017003992.1 | 2 | 854 | XP_016859481.1 | |
AK290906.1 | 3 | 819 | ||
AK312664.1 | 3 | 812 | ||
BC037961.1 | 3 | 875 | AAH37961.2 | |
L19593.1 | 3 | 911 | AAB59437.1 | |
M73969.1 | 1 | 528 | AAA83148.1 | |
M94582.1 | 2 | 555 | AAA36108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv961560 | Chr.2:218134916 - 218135916 on Build GRCh38 | Duplication | CXCR2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |