Assay Details
Target Gene Details
Entrez Gene ID: | 3275 |
Gene Name: | protein arginine methyltransferase 2 |
Gene Aliases: |
HRMT1L1 |
Location: |
Chr.21:46635153-46665685 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain | DIP2A S100B PRMT2 |
nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain | DIP2A S100B PRMT2 |
nsv588002 | Chr.21:46575530 - 46653004 on Build GRCh38 | Gain | S100B PRMT2 |
dgv4447n100 | Chr.21:46616138 - 46654452 on Build GRCh38 | Loss | PRMT2 |
esv34393 | Chr.21:46595759 - 46657900 on Build GRCh38 | Gain | S100B PRMT2 |
esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain | S100B PRMT2 |
esv2758822 | Chr.21:46566757 - 46657900 on Build GRCh38 | Gain | DIP2A S100B PRMT2 |
dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain | DIP2A S100B PRMT2 |
dgv4448n100 | Chr.21:46620242 - 46677033 on Build GRCh38 | Gain | PRMT2 |
esv3647172 | Chr.21:46592879 - 46650385 on Build GRCh38 | Loss | S100B PRMT2 |
nsv9865 | Chr.21:46648026 - 46652423 on Build GRCh38 | Gain | PRMT2 |
dgv4444n100 | Chr.21:46599898 - 46677033 on Build GRCh38 | Gain | S100B PRMT2 |
nsv1059706 | Chr.21:46627762 - 46662768 on Build GRCh38 | Gain | PRMT2 |
nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain | LOC101928796 C21orf58 PCBP3 SPATC1L DIP2A-IT1 YBEY FTCD S100B PRMT2 COL6A1 PCNT MCM3AP-AS1 DIP2A LSS MCM3AP COL6A2 |
dgv4449n100 | Chr.21:46634852 - 46677033 on Build GRCh38 | Gain+Loss | PRMT2 |
dgv4446n100 | Chr.21:46601742 - 46654452 on Build GRCh38 | Gain | S100B PRMT2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |