Assay Details
Target Gene Details
Entrez Gene ID: | 64928 |
Gene Name: | mitochondrial ribosomal protein L14 |
Gene Aliases: |
L14mt, L32mt, MRP-L14, MRP-L32, MRPL32, RMPL32, RPML32 |
Location: |
Chr.6:44113454-44127491 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL14 | NM_001318767.1 | 3 | 302 | NP_001305696.1 |
NM_001318768.1 | 3 | 246 | NP_001305697.1 | |
NM_001318769.1 | 3 | 235 | NP_001305698.1 | |
NM_001318770.1 | 3 | 508 | NP_001305699.1 | |
NM_001318771.1 | 2 | 252 | NP_001305700.1 | |
NM_032111.3 | 3 | 341 | NP_115487.2 | |
AA887574.1 | ||||
AK312086.1 | 3 | 273 | ||
BC065005.1 | 2 | 197 | AAH65005.1 | |
BG545603.1 | 3 | 506 | ||
BM013002.1 | 2 | 167 | ||
BU597168.1 | 3 | 268 | ||
BU753526.1 | 1 | 429 | ||
CF541260.1 | 1 | 430 | ||
CK823336.1 | 3 | 246 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462935 | Chr.6:44078406 - 44149870 on Build GRCh38 | Loss | TMEM63B MRPL14 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |