Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | NP_064536.2 | ||
XM_011511499.1 | XP_011509801.1 | |||
AF270647.1 | AAF81750.1 | |||
AK023234.1 | ||||
AK054615.1 | ||||
AK291190.1 | ||||
AL542720.3 | ||||
BC007894.2 | AAH07894.1 | |||
BC008894.2 | AAH08894.1 | |||
BC009998.2 | AAH09998.1 | |||
BC012774.2 | AAH12774.1 | |||
BC012793.2 | AAH12793.1 | |||
BC013279.2 | AAH13279.1 | |||
BC014076.2 | AAH14076.1 | |||
BC014274.2 | AAH14274.3 | |||
BC032106.1 | AAH32106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | ITPRIPL1 TMEM127 SNRNP200 LOC105373495 STARD7 CIAO1 NCAPH STARD7-AS1 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | CNNM4 ANKRD36C ANKRD36 STARD7 DUSP2 ASTL MIR3127 FER1L5 CNNM3 ITPRIPL1 ANKRD23 FAM178B LOC653924 LOC105373495 GPAT2 NCAPH LMAN2L STARD7-AS1 FAHD2B LOC101927053 LOC100506076 NEURL3 ADRA2B TMEM127 KANSL3 SEMA4C LOC105373496 ANKRD36B ANKRD39 LOC100506123 ARID5A SNRNP200 LOC107984110 CIAO1 FAHD2CP |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | CNNM4 ANKRD36C ANKRD36 STARD7 DUSP2 ASTL MIR3127 FER1L5 CNNM3 ITPRIPL1 ANKRD23 FAM178B LOC653924 LOC105373495 GPAT2 NCAPH LMAN2L STARD7-AS1 FAHD2B LOC101927053 LOC100506076 NEURL3 ADRA2B TMEM127 KANSL3 SEMA4C LOC105373496 ANKRD36B ANKRD39 LOC100506123 ARID5A SNRNP200 LOC107984110 CIAO1 FAHD2CP |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | CNNM4 ANKRD36C ANKRD36 STARD7 DUSP2 ASTL MIR3127 FER1L5 CNNM3 ITPRIPL1 ANKRD23 FAM178B LOC653924 LOC105373495 GPAT2 NCAPH LMAN2L STARD7-AS1 FAHD2B LOC101927053 LOC100506076 NEURL3 ADRA2B TMEM127 KANSL3 SEMA4C LOC105373496 ANKRD36B ANKRD39 LOC100506123 ARID5A SNRNP200 LOC107984110 CIAO1 FAHD2CP |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |