Assay Details
Target Gene Details
Entrez Gene ID: | 2997 |
Gene Name: | glycogen synthase 1 |
Gene Aliases: |
GSY, GYS |
Location: |
Chr.19:48968125-48993353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GYS1 | NM_001161587.1 | NP_001155059.1 | ||
NM_002103.4 | NP_002094.2 | |||
NR_027763.1 | ||||
AK223579.1 | BAD97299.1 | |||
AK291672.1 | ||||
AK294349.1 | ||||
AK300369.1 | ||||
AK302738.1 | ||||
AK303712.1 | ||||
BC002617.2 | AAH02617.1 | |||
BC003182.1 | AAH03182.1 | |||
BC007688.2 | AAH07688.1 | |||
J04501.1 | AAA88046.1 | |||
U32573.1 | AAB60385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | NTF4 SNAR-C4 CABP5 SNORD33 GFY SNAR-A7 ZNF114 LIG1 ZNF473 RRAS ADM5 C19orf73 SNAR-A5 VRK3 TSKS PLA2G4C LOC101928295 RCN3 TEAD2 PNKP CGB7 KDELR1 ALDH16A1 MIR6800 MIR6799 RPS11 FLT3LG SNAR-A3 C19orf68 SNORD35B CPT1C FUT2 GRWD1 SLC17A7 PTOV1-AS2 LHB MIR4750 TRPM4 SNRNP70 CARD8 SLC6A16 SNAR-A11 LOC105447645 PLEKHA4 TULP2 PTOV1-AS1 GRIN2D HSD17B14 RPL18 CA11 CCDC114 SNORD35A PRR12 PIH1D1 SNORD34 SNAR-A13 FUT1 CYTH2 SPHK2 RPL13A MIR4324 PPFIA3 SNAR-C5 PPP1R15A SNAR-A10 PRRG2 CD37 PTH2 MAMSTR CGB8 NUCB1 BAX BCAT2 LIN7B IL4I1 EMP3 LOC105372430 AKT1S1 SNAR-B2 CCDC155 SIGLEC11 GYS1 PLA2G4C-AS1 NTN5 FLJ26850 TMEM143 CGB5 ELSPBP1 FUZ DBP SNAR-G2 HRC SNAR-A12 CGB2 PTOV1 FTL SIGLEC16 LOC100287477 BCL2L12 MIR4751 RASIP1 LOC101059948 MIR150 SNAR-B1 KCNA7 SYNGR4 KCNJ14 MIR4749 SNAR-A1 MED25 SNAR-C2 CARD8-AS1 SNAR-A8 IRF3 MIR6798 NUCB1-AS1 SNAR-C3 FGF21 FCGRT DHDH SNAR-C1 SPACA4 RUVBL2 SNAR-A6 SNAR-A2 SNAR-A4 TBC1D17 SCAF1 CGB3 SNAR-A14 BSPH1 SNAR-A9 SNORD32A SEC1P NOSIP SULT2B1 ATF5 AP2A1 IZUMO1 LMTK3 NUP62 CGB1 SNAR-G1 PRMT1 MIR5088 FAM83E DKKL1 SNAR-D |
dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain | RUVBL2 LOC101059948 MIR6798 GYS1 LHB BAX FTL |
nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss | RUVBL2 NTF4 LOC101059948 GYS1 CGB7 LHB KCNA7 SNRNP70 CGB5 CGB3 PPFIA3 CGB1 SNAR-G1 C19orf73 MIR6798 NUCB1-AS1 CGB8 NUCB1 BAX SNAR-G2 LIN7B DHDH CGB2 FTL |
nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion | RUVBL2 NTF4 LOC101059948 GYS1 CGB7 LHB KCNA7 SNRNP70 CGB5 CGB3 CGB1 SNAR-G1 MIR6798 CGB8 BAX SNAR-G2 CGB2 FTL |
nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss | PLEKHA4 TULP2 PPP1R15A GYS1 NUCB1-AS1 NUCB1 BAX DHDH FTL |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |