Assay Details
Target Gene Details
Entrez Gene ID: | 1236 |
Gene Name: | C-C motif chemokine receptor 7 |
Gene Aliases: |
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1 |
Location: |
Chr.17:40553769-40565484 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCR7 | NM_001301714.1 | 2 | 1098 | NP_001288643.1 |
NM_001301716.1 | 3 | 1398 | NP_001288645.1 | |
NM_001301717.1 | 3 | 1154 | NP_001288646.1 | |
NM_001301718.1 | 3 | 1244 | NP_001288647.1 | |
NM_001838.3 | 3 | 1148 | NP_001829.1 | |
AK127810.1 | 3 | 1136 | ||
AK310277.1 | 3 | 1398 | ||
AK312732.1 | 3 | 1132 | ||
AY587876.1 | 2 | 1073 | AAT52232.1 | |
BC035343.1 | 3 | 1114 | AAH35343.1 | |
L08176.1 | 3 | 1136 | AAA58615.1 | |
L31581.1 | 3 | 1139 | AAA74231.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |