Assay Details
Target Gene Details
Entrez Gene ID: | 100506012 |
Gene Name: | PPP5 tetratricopeptide repeat domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.19:46480788-46601200 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP5D1 | NM_001205281.1 | NP_001192210.1 | ||
BM718195.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |