Assay Details
Target Gene Details
Entrez Gene ID: | 417 |
Gene Name: | ADP-ribosyltransferase 1 |
Gene Aliases: |
ART2, ARTC1, CD296, RT6 |
Location: |
Chr.11:3645110-3665595 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ART1 | NM_004314.2 | NP_004305.2 | ||
XM_011520114.2 | XP_011518416.1 | |||
XM_017017763.1 | XP_016873252.1 | |||
BC069102.1 | AAH69102.1 | |||
BC111729.1 | AAI11730.1 | |||
S74683.1 | AAB32387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759796 | Chr.11:3215282 - 3780199 on Build GRCh38 | Gain+Loss | MRGPRG-AS1 MRGPRE ART1 LOC105376525 ART5 NUP98 TSSC2 CHRNA10 LOC101927708 MRGPRG OR7E12P TRPC2 LOC105376526 ZNF195 |
nsv526441 | Chr.11:3604947 - 3735456 on Build GRCh38 | Gain | ART1 ART5 NUP98 CHRNA10 TRPC2 |
nsv428247 | Chr.11:3595654 - 3780199 on Build GRCh38 | Loss | ART1 ART5 NUP98 CHRNA10 TRPC2 |
nsv832051 | Chr.11:3502717 - 3707484 on Build GRCh38 | Gain | ART1 ART5 NUP98 CHRNA10 LOC101927708 TRPC2 |
nsv1046356 | Chr.11:3607607 - 3752985 on Build GRCh38 | Gain | ART1 ART5 NUP98 CHRNA10 TRPC2 |
esv3625147 | Chr.11:3607625 - 3732043 on Build GRCh38 | Gain | ART1 ART5 NUP98 CHRNA10 TRPC2 |
esv3625148 | Chr.11:3609614 - 3680474 on Build GRCh38 | Gain | ART1 ART5 NUP98 CHRNA10 TRPC2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |