Assay Details
Target Gene Details
Entrez Gene ID: | 153769 |
Gene Name: | SH3 domain containing ring finger 2 |
Gene Aliases: |
HEPP1, POSHER, PPP1R39, RNF158 |
Location: |
Chr.5:145936563-146081773 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3RF2 | NM_152550.3 | 2 | 437 | NP_689763.3 |
XM_005268380.2 | 2 | 433 | XP_005268437.1 | |
XM_006714757.1 | 2 | 433 | XP_006714820.1 | |
XM_011537567.2 | 2 | 433 | XP_011535869.1 | |
XM_011537568.2 | 2 | 427 | XP_011535870.1 | |
XM_011537569.2 | 2 | 433 | XP_011535871.1 | |
XM_017009136.1 | 2 | 427 | XP_016864625.1 | |
AK074234.1 | 2 | 421 | BAB85025.1 | |
AK292576.1 | 2 | 422 | ||
AL833297.1 | 1 | 251 | ||
BC125106.1 | 2 | 394 | ||
BC125107.1 | 2 | 394 | ||
BP352041.1 | 2 | 440 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |