Assay Details
Target Gene Details
Entrez Gene ID: | 23387 |
Gene Name: | SIK family kinase 3 |
Gene Aliases: |
L19, QSK, SIK-3 |
Location: |
Chr.11:116843402-117098453 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIK3 | NM_001281748.1 | 20 | 2455 | NP_001268677.1 |
NM_001281749.1 | 19 | 2331 | NP_001268678.1 | |
NM_025164.4 | 19 | 2331 | NP_079440.3 | |
XM_005271481.2 | 20 | 2513 | XP_005271538.1 | |
XM_005271482.4 | 19 | 2369 | XP_005271539.2 | |
XM_005271484.3 | 19 | 2369 | XP_005271541.2 | |
XM_005271485.3 | 7 | 851 | XP_005271542.1 | |
XM_011542721.2 | 20 | 2475 | XP_011541023.1 | |
XM_011542722.1 | 20 | 2513 | XP_011541024.1 | |
XM_011542723.2 | 20 | 2356 | XP_011541025.1 | |
XM_011542724.2 | 19 | 2210 | XP_011541026.1 | |
XM_011542725.2 | 18 | 2093 | XP_011541027.1 | |
XM_017017424.1 | 18 | 2225 | XP_016872913.1 | |
XM_017017425.1 | 20 | 2551 | XP_016872914.1 | |
XM_017017426.1 | 18 | 2425 | XP_016872915.1 | |
XM_017017427.1 | 15 | 1828 | XP_016872916.1 | |
AB209327.1 | 12 | 1341 | BAD92564.1 | |
AK122838.1 | 5 | 1773 | ||
BC035583.2 | 2 | 161 | AAH35583.3 | |
BC063887.1 | 5 | 524 | AAH63887.1 | |
BC128510.1 | 19 | 2193 | ||
BC128511.1 | 20 | 2311 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |