Assay Details
Target Gene Details
Entrez Gene ID: | 7701 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
pHZ-49 |
Location: |
Chr.2:218637592-218659632 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF142 | NM_001105537.2 | 8 | 3544 | NP_001099007.1 |
NR_073599.1 | 9 | 3853 | ||
NR_073600.1 | 9 | 3686 | ||
XM_006712730.3 | 9 | 4219 | XP_006712793.1 | |
XM_011511784.2 | 9 | 4314 | XP_011510086.1 | |
XM_011511785.2 | 9 | 4239 | XP_011510087.1 | |
XM_011511786.2 | 9 | 4144 | XP_011510088.1 | |
XM_011511787.2 | 8 | 4139 | XP_011510089.1 | |
XM_011511788.2 | 8 | 4044 | XP_011510090.1 | |
XM_011511789.2 | 8 | 3712 | XP_011510091.1 | |
XM_017004871.1 | 8 | 4295 | XP_016860360.1 | |
XM_017004872.1 | 7 | 3695 | XP_016860361.1 | |
AK095027.1 | ||||
D87073.1 | 8 | 3542 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3594346 | Chr.2:218640692 - 218643860 on Build GRCh38 | Loss | ZNF142 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |