Assay Details
Target Gene Details
Entrez Gene ID: | 28952 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
CXorf37, JM1, RTSC2 |
Location: |
Chr.X:49235467-49250526 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC22 | NM_014008.4 | NP_054727.1 | ||
XM_005272599.3 | XP_005272656.1 | |||
AJ005890.1 | CAA06747.1 | |||
AK291976.1 | ||||
AK296911.1 | 7 | 1014 | ||
AK298264.1 | ||||
AK316375.1 | ||||
BC000972.2 | AAH00972.1 | |||
BC011675.2 | AAH11675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | FOXP3 CACNA1F PPP1R3F CCDC22 GAGE10 |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | SLC35A2 FOXP3 LOC101927635 PPP1R3F KCND1 WAS SLC38A5 GLOD5 PORCN PCSK1N WDR45 FTSJ1 SUV39H1 CCDC120 PIM2 PLP2 SYP-AS1 PRAF2 CCDC22 WDR13 GPKOW EBP PRICKLE3 SYP TFE3 RBM3 GATA1 PQBP1 TBC1D25 MAGIX HDAC6 OTUD5 TIMM17B CACNA1F ERAS GRIPAP1 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SLC35A2 ZNF630-AS1 FOXP3 ZNF630 SSX9 LOC101927635 PPP1R3F KCND1 WAS SLC38A5 GLOD5 PORCN PCSK1N WDR45 SSX3 FTSJ1 SUV39H1 ZNF182 CCDC120 PIM2 PLP2 SYP-AS1 PRAF2 CCDC22 SPACA5 SSX5 WDR13 GPKOW SSX4 SSX1 EBP SSX4B PRICKLE3 SPACA5B SYP TFE3 RBM3 GATA1 SSX6 PQBP1 TBC1D25 MAGIX HDAC6 OTUD5 TIMM17B CACNA1F ERAS GRIPAP1 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | FOXP3 CACNA1F PPP1R3F CCDC22 GAGE10 |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | FOXP3 CACNA1F CCDC22 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |