Assay Details
Target Gene Details
Entrez Gene ID: | 64794 |
Gene Name: | DEAD-box helicase 31 |
Gene Aliases: |
PPP1R25 |
Location: |
Chr.9:132592997-132670403 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 25 - Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX31 | NM_001322340.1 | NP_001309269.1 | ||
NM_001322341.1 | NP_001309270.1 | |||
NM_001322342.1 | NP_001309271.1 | |||
NM_001322343.1 | NP_001309272.1 | |||
NM_022779.8 | NP_073616.6 | |||
NR_136309.1 | ||||
XM_005272207.4 | XP_005272264.1 | |||
XM_006717236.3 | XP_006717299.1 | |||
XM_011518921.2 | XP_011517223.1 | |||
XM_011518922.2 | XP_011517224.1 | |||
AF335568.1 | AAQ14889.1 | |||
AF335569.1 | AAQ14890.1 | |||
AF427339.1 | AAL26549.1 | |||
AK023695.1 | BAB14644.1 | |||
AK027002.1 | BAB15620.1 | |||
AK225121.1 | ||||
AK225350.1 | ||||
AK302670.1 | ||||
BC012726.2 | AAH12726.2 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |