Assay Details
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMPD3 | NM_018667.3 | 8 | 2237 | NP_061137.1 |
XM_005256031.3 | 8 | 2330 | XP_005256088.1 | |
XM_005256032.3 | 8 | 2349 | XP_005256089.1 | |
XM_011523207.1 | 9 | 2330 | XP_011521509.1 | |
XM_011523208.2 | 7 | 2431 | XP_011521510.1 | |
XM_011523209.2 | 7 | 2214 | XP_011521511.1 | |
XM_017023405.1 | 8 | 2156 | XP_016878894.1 | |
XM_017023406.1 | 9 | 2637 | XP_016878895.1 | |
XM_017023407.1 | 9 | 2359 | XP_016878896.1 | |
XM_017023408.1 | 9 | 2313 | XP_016878897.1 | |
AJ250460.1 | 6 | 1815 | CAB92964.1 | |
AK289651.1 | 8 | 2238 | ||
BC112238.1 | 6 | 1892 | AAI12239.1 | |
BC143631.1 | 6 | 1868 | ||
BC143634.1 | 6 | 1841 | ||
DQ424899.1 | 6 | 1764 | ABD83664.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |