Assay Details
Target Gene Details
Entrez Gene ID: | 2896 |
Gene Name: | granulin |
Gene Aliases: |
CLN11, GEP, GP88, PCDGF, PEPI, PGRN |
Location: |
Chr.17:44345086-44353106 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRN | NM_002087.3 | 13 | 2016 | NP_002078.1 |
XM_005257253.1 | 13 | 2061 | XP_005257310.1 | |
AF055008.1 | 13 | 1822 | AAC09359.1 | |
AK000607.1 | 13 | 1800 | ||
AK023348.1 | 10 | 1304 | BAB14535.1 | |
AK222522.1 | 13 | 1800 | BAD96242.1 | |
AK296090.1 | 13 | 1754 | ||
AK303830.1 | 10 | 1326 | ||
AY124489.1 | 12 | 1778 | AAM94026.1 | |
BC000324.2 | 12 | 1514 | AAH00324.1 | |
BC010577.2 | 12 | 1770 | AAH10577.1 | |
BT006844.1 | AAP35490.1 | |||
M75161.1 | 12 | 1772 | AAA58617.1 | |
X62320.1 | 13 | 1800 | CAA44196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5606n54 | Chr.17:44350262 - 44355375 on Build GRCh38 | Loss | FAM171A2 GRN |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |