Assay Details
Target Gene Details
Entrez Gene ID: | 662 |
Gene Name: | BCL2 interacting protein 1 |
Gene Aliases: |
NIP1, SEC20, TRG-8 |
Location: |
Chr.5:173144442-173164387 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BNIP1 | NM_001205.2 | 6 | 964 | NP_001196.2 |
NM_013978.2 | 5 | 862 | NP_053581.2 | |
NM_013979.2 | 7 | 1093 | NP_053582.2 | |
NM_013980.2 | 6 | 991 | NP_053583.2 | |
XM_011534638.1 | 8 | 1161 | XP_011532940.1 | |
AF083956.1 | AAC33124.1 | |||
AF083957.1 | 7 | 999 | AAC33125.1 | |
AF083958.1 | 6 | 897 | AAC33126.1 | |
BC010959.1 | 6 | 889 | AAH10959.1 | |
BP384034.1 | ||||
U15172.1 | 6 | 870 | AAC00020.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |