Assay Details
Target Gene Details
Entrez Gene ID: | 7009 |
Gene Name: | transmembrane BAX inhibitor motif containing 6 |
Gene Aliases: |
BAXI1, BI-1, TEGT |
Location: |
Chr.12:49741510-49764934 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMBIM6 | NM_001098576.1 | 10 | 2719 | NP_001092046.1 |
NM_003217.2 | 10 | 2349 | NP_003208.2 | |
XM_005269126.3 | 11 | 2621 | XP_005269183.1 | |
AK026605.1 | 1 | 1102 | ||
AK124849.1 | ||||
AK130050.1 | 10 | 2302 | ||
AK130462.1 | 8 | 2171 | ||
AK225804.1 | 10 | 2301 | ||
BC000916.1 | 10 | 2282 | AAH00916.1 | |
BC036203.1 | 10 | 2357 | ||
BQ613936.1 | 1 | 326 | ||
X75861.1 | 9 | 2257 | CAA53472.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |