Assay Details
Target Gene Details
Entrez Gene ID: | 84913 |
Gene Name: | atonal bHLH transcription factor 8 |
Gene Aliases: |
HATH6, bHLHa21 |
Location: |
Chr.2:85753786-85791383 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATOH8 | NM_032827.6 | 1 | 486 | NP_116216.2 |
XM_006712122.3 | 1 | 486 | XP_006712185.1 | |
XM_011533139.1 | 1 | 486 | XP_011531441.1 | |
AF529205.1 | 1 | 82 | AAO85773.1 | |
AK027614.1 | 1 | 279 | BAB55233.1 | |
AK074681.1 | 1 | 378 | BAC11135.1 | |
AK096049.1 | ||||
BC021207.2 | 1 | 377 | AAH21207.1 | |
BC094832.1 | 1 | 156 | AAH94832.1 | |
DA629171.1 | 1 | 486 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829487 | Chr.2:85753476 - 85755998 on Build GRCh38 | Gain | ATOH8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |