Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AF218026.1 | AAG17268.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AY358168.1 | AAQ88535.1 | |||
BC015172.1 | AAH15172.1 | |||
U79287.1 |
Target Gene Details
Entrez Gene ID: | 101928378 |
Gene Name: | PTOV1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.19:49856465-49859289 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1-AS2 | NR_110730.1 | |||
AL050131.1 | 1 | 608 | ||
CK903988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)