Assay Details
Target Gene Details
Entrez Gene ID: | 84899 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.13:100603836-100675025 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMTC4 | NM_001079669.2 | 11 | 1645 | NP_001073137.1 |
NM_001286453.1 | 9 | 1312 | NP_001273382.1 | |
NM_032813.3 | 12 | 1855 | NP_116202.2 | |
XM_011521121.2 | 13 | 2007 | XP_011519423.1 | |
XM_011521122.2 | 13 | 1923 | XP_011519424.1 | |
XM_011521123.2 | 13 | 2084 | XP_011519425.1 | |
XM_011521124.2 | 12 | 1712 | XP_011519426.1 | |
XM_011521125.1 | 13 | 1916 | XP_011519427.1 | |
XM_017020796.1 | 12 | 1910 | XP_016876285.1 | |
XM_017020797.1 | 11 | 1649 | XP_016876286.1 | |
XM_017020798.1 | 11 | 1745 | XP_016876287.1 | |
XM_017020799.1 | 10 | 1430 | XP_016876288.1 | |
AF339792.1 | 9 | 1227 | ||
AK027530.1 | 6 | 738 | BAB55179.1 | |
AK056409.1 | 12 | 1714 | ||
AK092653.1 | 4 | 1728 | ||
AK290345.1 | 11 | 1505 | ||
AK299859.1 | 9 | 1151 | ||
BC018707.1 | 6 | 816 | AAH18707.1 | |
BG830268.1 | 4 | 531 | ||
BX647956.1 | 11 | 1536 | ||
BX648008.1 | 11 | 1489 |