Assay Details
Target Gene Details
Entrez Gene ID: | 64223 |
Gene Name: | MTOR associated protein, LST8 homolog |
Gene Aliases: |
GBL, GbetaL, LST8, POP3, WAT1 |
Location: |
Chr.16:2205177-2209417 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLST8 | NM_001199173.1 | NP_001186102.1 | ||
NM_001199174.1 | NP_001186103.1 | |||
NM_001199175.1 | NP_001186104.1 | |||
NM_022372.4 | NP_071767.3 | |||
XM_005255475.3 | XP_005255532.2 | |||
XM_005255478.2 | XP_005255535.1 | |||
XM_005255479.2 | XP_005255536.1 | |||
XM_017023548.1 | XP_016879037.1 | |||
XM_017023549.1 | XP_016879038.1 | |||
XM_017023550.1 | XP_016879039.1 | |||
XM_017023551.1 | 5 | 774 | XP_016879040.1 | |
XM_017023552.1 | XP_016879041.1 | |||
AF195883.1 | AAF04308.1 | |||
AK021536.1 | ||||
AK022227.1 | BAB13990.1 | |||
AK098762.1 | ||||
AK302201.1 | ||||
AU121635.1 | ||||
AY223837.1 | AAO73410.1 | |||
BC001313.2 | AAH01313.1 | |||
BC017119.1 | AAH17119.1 | |||
BC052292.1 | AAH52292.1 | |||
BC088354.1 | AAH88354.1 | |||
BI761765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)