Assay Details
Target Gene Details
Entrez Gene ID: | 10768 |
Gene Name: | adenosylhomocysteinase like 1 |
Gene Aliases: |
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA |
Location: |
Chr.1:109984741-110023742 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHCYL1 | NM_001242673.1 | NP_001229602.1 | ||
NM_001242674.1 | NP_001229603.1 | |||
NM_001242675.1 | NP_001229604.1 | |||
NM_001242676.1 | NP_001229605.1 | |||
NM_006621.5 | NP_006612.2 | |||
XM_011540535.1 | XP_011538837.1 | |||
AF315687.1 | AAL26869.1 | |||
AK131563.1 | 1 | 724 | BAD18696.1 | |
AK303690.1 | ||||
AK308080.1 | ||||
AK316110.1 | ||||
AL049954.1 | CAB43223.2 | |||
BC007576.1 | ||||
BC010681.1 | ||||
BC016942.1 | ||||
BC065254.1 | ||||
BC095476.1 | ||||
BC110896.1 | ||||
BC111563.1 | AAI11564.1 | |||
U82761.1 | AAC01960.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |