Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA25 | NM_024953.3 | 16 | 1885 | NP_079229.2 |
XM_005253938.1 | 15 | 1731 | XP_005253995.1 | |
XM_006719606.2 | 16 | 2545 | XP_006719669.1 | |
XM_017019977.1 | 14 | 1601 | XP_016875466.1 | |
AB054990.1 | 16 | 1827 | BAC80174.1 | |
AK023151.1 | 13 | 1530 | BAB14432.1 | |
AK292485.1 | 16 | 1858 | ||
AK307494.1 | 17 | 2034 | ||
AK309772.1 | 16 | 1840 | ||
BC034357.1 | 16 | 1839 | ||
BC113585.1 | 16 | 1885 | ||
BC113587.1 | 16 | 1885 | ||
BX641140.1 | 16 | 1840 | CAE46062.1 | |
BX647450.1 | 16 | 1838 | ||
BX647992.1 | 14 | 1615 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |