Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | ||||
DA409636.1 | ||||
DB096906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
dgv3982n54 | Chr.14:105167384 - 105178045 on Build GRCh38 | Loss | NUDT14 JAG2 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | LOC100507437 BTBD6 MIR6765 NUDT14 JAG2 PACS2 LOC102723354 MTA1 GPR132 BRF1 CRIP2 TEX22 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | IGH NUDT14 JAG2 MTA1 C14orf80 BRF1 CRIP2 LOC100507437 IGHA2 BTBD6 LOC105370697 MIR6765 TMEM121 PACS2 CRIP1 TEX22 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | MIR6765 NUDT14 JAG2 LOC102723354 GPR132 BRF1 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | MIR6765 NUDT14 JAG2 BRF1 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | C14orf180 LOC388022 CDCA4 AKT1 LOC101929634 MIR203B APOPT1 LOC105370687 C14orf79 AHNAK2 BAG5 XRCC3 BRF1 PLD4 TRMT61A INF2 SNORA28 BTBD6 ZFYVE21 ZBTB42 LINC00638 TMEM179 MIR6765 CKB TDRD9 SIVA1 C14orf2 MARK3 CEP170B GPR132 LINC00637 LOC105378183 ADSSL1 KIF26A KLC1 NUDT14 JAG2 ASPG EIF5 MIR4710 LOC105370708 PPP1R13B LOC105370691 LOC107984648 RD3L PACS2 LOC102723354 MIR203A |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | BTBD6 CDCA4 MIR6765 NUDT14 JAG2 LOC102723354 C14orf79 AHNAK2 GPR132 BRF1 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHD IGHM MIR4539 IGHV3-30 C14orf80 BRF1 MIR8071-2 MIR8071-1 BTBD6 LOC105370697 MIR4537 IGHG3 MIR6765 IGHV4-55 IGHV3-69-1 CRIP1 IGHG2 IGHA1 ELK2AP TEX22 IGHV1-69 IGHG4 IGH ADAM6 IGHV3-48 IGHV3-41 NUDT14 JAG2 IGHV3-23 IGHG1 MTA1 IGHV3-71 CRIP2 LOC100507437 MIR4507 IGHA2 LINC00226 LOC107987211 MIR4538 LINC00221 KIAA0125 TMEM121 PACS2 IGHV4-31 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | CDCA4 MIR6765 NUDT14 JAG2 LOC102723354 C14orf79 AHNAK2 GPR132 BRF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |