Assay Details
Target Gene Details
Entrez Gene ID: | 9044 |
Gene Name: | B-TFIID TATA-box binding protein associated factor 1 |
Gene Aliases: |
MOT1, TAF(II)170, TAF172, TAFII170 |
Location: |
Chr.10:91923769-92030998 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BTAF1 | NM_003972.2 | 4 | 437 | NP_003963.1 |
XM_011540326.1 | 4 | 647 | XP_011538628.1 | |
XM_011540329.2 | 4 | 647 | XP_011538631.1 | |
XM_017016877.1 | XP_016872366.1 | |||
XM_017016878.1 | 3 | 325 | XP_016872367.1 | |
XM_017016879.1 | 3 | 325 | XP_016872368.1 | |
XM_017016880.1 | 3 | 325 | XP_016872369.1 | |
XM_017016881.1 | XP_016872370.1 | |||
AF038362.1 | 4 | 456 | AAC04573.1 | |
AJ001017.2 | 4 | 351 | CAA04475.1 | |
AK308399.1 | 4 | 647 | ||
BC112201.1 | 4 | 421 | AAI12202.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |