Assay Details
Target Gene Details
Entrez Gene ID: | 116173 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 5 |
Gene Aliases: |
CKLFSF5 |
Location: |
Chr.14:23376439-23379772 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM5 | NM_001037288.1 | 1 | 330 | NP_001032365.1 |
NM_001288744.1 | 1 | 330 | NP_001275673.1 | |
NM_001288745.1 | 1 | 330 | NP_001275674.1 | |
NM_001288746.1 | 1 | 330 | NP_001275675.1 | |
NM_138460.2 | 1 | 330 | NP_612469.1 | |
XM_017020953.1 | XP_016876442.1 | |||
XM_017020954.1 | XP_016876443.1 | |||
AF479262.2 | 1 | 331 | AAN73040.1 | |
AF527413.1 | 1 | 113 | AAQ08985.1 | |
AF527414.1 | AAQ08986.1 | |||
AF527948.2 | 1 | 101 | AAQ09008.2 | |
AF527949.1 | 1 | 101 | AAQ09009.1 | |
AK094840.1 | 1 | 91 | BAC04432.1 | |
AK289886.1 | 1 | 85 | ||
BC013109.1 | AAH13109.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |